missing translation for 'onlineSavingsMsg'
Learn More

D-64131, Tocris Bioscience™

Tubulin polymerization inhibitor. Antitumor in vivo

Brand:  Tocris (Bio-Techne) 164350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449853

  • £504.21 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Novel inhibitor of tubulin polymerization; cytotoxic and inhibits tumor cell proliferation in vitro (IC50 = 74 nM). Prevents growth of tumor models in mice following oral administration in vivo.
Specifications

Chemical Identifiers

74588-78-6
251.285
5-methoxy-1h-indol-2-yl phenylmethanone, 5-methoxy-1h-indol-2-yl-phenylmethanone, 5-methoxy-1h-indol-2-yl phenyl methanone, 5-methoxy-1h-indol-2-yl-phenyl-methanone, 2-benzoyl-5-methoxy-1h-indole, tocris-1643, 5-methoxy-2-benzoylindole, 2-benzoyl-5-methoxyindole
(5-methoxy-1H-indol-2-yl)-phenylmethanone
C16H13NO2
ICMIJSRDISNKOC-UHFFFAOYSA-N
3921152
COC1=CC2=C(C=C1)NC(=C2)C(=O)C3=CC=CC=C3

Specifications

50mg
74588-78-6
5-methoxy-1h-indol-2-yl phenylmethanone, 5-methoxy-1h-indol-2-yl-phenylmethanone, 5-methoxy-1h-indol-2-yl phenyl methanone, 5-methoxy-1h-indol-2-yl-phenyl-methanone, 2-benzoyl-5-methoxy-1h-indole, tocris-1643, 5-methoxy-2-benzoylindole, 2-benzoyl-5-methoxyindole
COC1=CC2=C(C=C1)NC(=C2)C(=O)C3=CC=CC=C3
251.285
251.28
D-64131
C16H13NO2
ICMIJSRDISNKOC-UHFFFAOYSA-N
(5-methoxy-1H-indol-2-yl)-phenylmethanone
3921152
>99%
Videos
Documents