missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%
£53.24 - £206.00
Chemical Identifiers
CAS | 17609-48-2 |
---|---|
Molecular Formula | C10H14ClNO2 |
Molecular Weight (g/mol) | 215.677 |
MDL Number | MFCD00052005 |
InChI Key | FNNXQLSKQSVNLL-SBSPUUFOSA-N |
Synonym | r-ethyl 2-amino-2-phenylacetate hydrochloride, r-ethyl 2-amino-2-phenylacetate hcl, ethyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine-oet hcl, h-d-phg-oet . hcl, d---2-phenylglycine ethyl ester hydrochloride, d---alpha-phenylglycine ethyl ester hydrochloride, pubchem10931, d-phg-oet hydrochloride |
PubChem CID | 20833765 |
IUPAC Name | ethyl (2R)-2-amino-2-phenylacetate;hydrochloride |
SMILES | CCOC(=O)C(C1=CC=CC=C1)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381327
|
Alfa Aesar™
L01564.09 |
10g |
£53.24
10g |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11391327
|
Alfa Aesar™
L01564.18 |
50g |
£206.00
50g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ApplicationsD-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
17609-48-2 | |
215.677 | |
FNNXQLSKQSVNLL-SBSPUUFOSA-N | |
20833765 | |
CCOC(=O)C(C1=CC=CC=C1)N.Cl |
C10H14ClNO2 | |
MFCD00052005 | |
r-ethyl 2-amino-2-phenylacetate hydrochloride, r-ethyl 2-amino-2-phenylacetate hcl, ethyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine-oet hcl, h-d-phg-oet . hcl, d---2-phenylglycine ethyl ester hydrochloride, d---alpha-phenylglycine ethyl ester hydrochloride, pubchem10931, d-phg-oet hydrochloride | |
ethyl (2R)-2-amino-2-phenylacetate;hydrochloride |
Specifications
D-(-)-2-Phenylglycine ethyl ester hydrochloride | |
C10H14ClNO2 | |
5775455 | |
Soluble in water. | |
CCOC(=O)C(C1=CC=CC=C1)N.Cl | |
215.677 | |
215.68 | |
∽190°C (decomposition) | |
−94° (c=1 in Water) |
17609-48-2 | |
MFCD00052005 | |
r-ethyl 2-amino-2-phenylacetate hydrochloride, r-ethyl 2-amino-2-phenylacetate hcl, ethyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine-oet hcl, h-d-phg-oet . hcl, d---2-phenylglycine ethyl ester hydrochloride, d---alpha-phenylglycine ethyl ester hydrochloride, pubchem10931, d-phg-oet hydrochloride | |
FNNXQLSKQSVNLL-SBSPUUFOSA-N | |
ethyl (2R)-2-amino-2-phenylacetate;hydrochloride | |
20833765 | |
≥98% | |
10g |