missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-2-Aminoadipic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 186°C to 188°C |
Color | White |
Elemental Analysis | 43.60 to 45.84% C, 8.47 to 8.91% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Rotation | -20.5 |
Formula Weight | 161.16 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'