missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 7, eFluor 615, clone: LP5K, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 42-9005-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381120

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

This LP5K monoclonal antibody reacts with human cytokeratin 7 (K7), a 54kDa type II (or basic) keratin expressed either alone or paired with cytokeratin 19 in simple epithelia, mesothelium, urothelium, and pseudostratified epithelium. Expression of cytokeratin 7 in the gastric foveolar, intestinal, and stratified squamous epithelia is extremely low or undetectable. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. In addition, most carcinomas express cytokeratin 7. The coordinated expression of this keratin with cytokeratin 20 is commonly used as a diagnostic marker for a variety of carcinomas.
Specifications

Specifications

Cytokeratin 7
LP5K
eFluor 615
Conjugated
IgG2b
RUO
Primary
Human
Immunohistochemistry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.