Cytokeratin 6 Mouse anti-Human, Clone: LL020, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 14-9788-80

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15527686

Quantity Price
1 £ 55.59 / 25µg
Estimated Shipment date
from Supplier 05-06-2017
Add to basket


Description and Specification

Specification

Antigen Cytokeratin 6
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Applications Western Blot
Clone LL020
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Host Species Murine
Isotype IgM
Quantity 25μg
Regulatory Status RUO
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

The monoclonal antibody LL020 recognizes human Cytokeratin 6. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. Cytokeratin 6 is a type II (basic) keratin that associates with Cytokeratin 16 and Cytokeratin 17. Cytokeratin 6 is expressed in the epidermis, outer root sheath of hair follicles, oral mucosa, suprabasal cells of stratified epithelia, and glandular epithelia. Gene mutations result in inherited skin and hair disorders. Some reports suggest that LL020 may have some crossreactivity to Cytokeratin 5.