missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568043
Additional Details : Weight : 0.01000kg
Description
Cytokeratin 34 is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.Specifications
Cytokeratin 34 | |
Polyclonal | |
Unconjugated | |
KRT34 | |
HA4; Ha-4; KRTHA4; hHa4; K34; hair keratin; type I Ha4; hard keratin; type I; 4; keratin; hair; acidic; 4; keratin; type I cuticular Ha4; keratin-34 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3885 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O76011 | |
KRT34 | |
Synthesized peptide derived from human Cytokeratin 34. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |