missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (KRT18/836) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48066
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Canine, Feline, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KRT18 | |
| Recombinant full-length human Cytokeratin 18 protein (Uniprot: P05783) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It is reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. | |
| Store at 4C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| KRT18/836 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Mouse | |
| 45 kDa | |
| 7 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers | |
| 3875 | |
| Human, Canine, Feline, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction