missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Mouse, Clone: SPM190, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3866-MSM1X-P1
Additional Details : Weight : 0.01000kg
Specifications
Cytokeratin 15 | |
Cytoplasmic | |
SPM190 | |
Unconjugated | |
NP_016880103 | |
Mouse | |
52kDa | |
Protein A/G Purified | |
RUO | |
3866 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
KRT15 | |
A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 | |
A431 cells. Skin or Colon | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |