Learn More
Thermo Scientific™ Cytochrome C, equine heart, 90+%
Cytochrome C, CAS # 9007-43-6, is a component of the electron transport chain in mitochondria, making it an important electron transporter that forms an electron bridge between respirable substrates and oxygen.
POA
Description
Description
A component of the electron transport chain in mitochondria, cytochrome c is assumed to be the functional complex utilized in low-level laser therapy (LLLT), which increases the metabolic activity and frees up more energy for the cells to repair the tissue.It shows peroxidase activity by oxidation of various electron donors such as 2,2-azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) (ABTS), 2-keto-4-thiomethyl butyric acid and 4-aminoantipyrine. It is used as an important mediator in apoptotic pathways.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General description
• Cytochrome C is a key player of the electron transport chain in mitochondria
• It plays an important role in apoptotic pathways
• It also plays a role in cellular oxidation in animals and plants
which make it an effective biological electron carrier.
Applications
• Cytochrome c shows peroxidase activity by oxidation of various electron donors such as 2,2-azino-bis (3-ethylbenzthiazoline-6-sulphonic acid) (ABTS), 2-keto-4-thiomethyl butyric acid and 4-aminoantipyrine
• It is assumed to be the functional complex utilized in low-level laser therapy (LLLT), which increases the metabolic activity and frees up more energy for the cells to repair the tissue
• It is used as an important mediator in apoptotic pathways