missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cytochrome C, Equine Heart, 90+%

For research use

Brand:  Alfa Aesar™ J62122.03

Code : NEW

Additional Details : CAS Number : 9007-43-6 Weight : 0.00100kg

 View more versions of this product

Product Code. 15454559

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only


Specifications

Specifications

A component of the electron transport chain in mitochondria
Lyophilized powder
1g
-30° to -10°C
Cytochrome C, Equine Heart, ≥90%
Lyophilized Powder
≥90%
Equine heart
RUO
No
Videos
SDS