missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cytochrome C, Equine Heart, 90+%

For research use

Brand:  Alfa Aesar™ J62122.MD

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9007-43-6 Weight : 0.00025kg

 View more versions of this product

Product Code. 15464559

  • POA / 250mg


Estimated Shipment date
from Supplier 07-08-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

A component of the electron transport chain in mitochondria
Lyophilized powder
250mg
-30° to -10°C
Cytochrome C, Equine Heart, ≥90%
Lyophilized Powder
≥90%
Equine heart
RUO
No
Documentation

Technical Documents