missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b5, rHuman
Cytochrome b5, rHuman
Brand: Invitrogen P2252
Code : 50
Additional Details : Weight : 3.36000kg
Description
Cytochrome b5 is a membrane-bound hemoprotein that enhances the catalytic efficiency of some P450 isoforms. Our human cytochrome b5 is overexpressed in E. coli and purified to homogeneity. This enzyme is provided in solution.For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
100 μg | |
Recombinant | |
Dry Ice | |
Toxicology & Drug Metabolism |
Human | |
Store at -80°C | |
E. coli |
For Research Use Only. Not for use in diagnostic procedures.