missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine, 99+%

Catalog Number 10182420
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
65-46-3
C9H13N3O5
243.22
MFCD00006545
UHDGCWIWMRVCDJ-UHFFFAOYNA-N
cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
6175
CHEBI:17562
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
65-46-3
C9H13N3O5
243.22
MFCD00006545
UHDGCWIWMRVCDJ-UHFFFAOYNA-N
cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
6175
CHEBI:17562
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
Melting Point 210.0°C to 217.0°C
Color White
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 31,24
Merck Index 15,2783
Specific Rotation + 31.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alkalies,insoluble in organic solvents
Spectrophotometry 98 to 102% (lambda max. 280nm,pH 1) molar absorptivity 13000+/-2%,on dry basis
Transmittance 98% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 243.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, 99.5%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'