missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-triphosphate, disodium salt hydrate, 95%, ACROS Organics™

£151.00

Chemical Identifiers

CAS 652154-13-7
Molecular Formula C9H14N3Na2O14P3·xH2O
Molecular Weight (g/mol) 527.12
MDL Number MFCD00150807
InChI Key LDTPLTXNLOEHLN-ODQFIEKDSA-N
Synonym cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
PubChem CID 129893565
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
View More Specs

Produits 1
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
10062840
FDS missing translation for 'viewDocuments'
Acros Organics
226225000
500mg
£151.00
500mg
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spécification

Identifiants chimiques

652154-13-7
527.12
LDTPLTXNLOEHLN-ODQFIEKDSA-N
129893565
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
C9H14N3Na2O14P3·xH2O
MFCD00150807
cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium

Spécification

Cytidine 5′-triphosphate,disodium salt hydrate
Glass bottle
White
500mg
652154-13-7
C9H14N3Na2O14P3·xH2O
cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
LDTPLTXNLOEHLN-ODQFIEKDSA-N
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
129893565
Crystalline Powder
Authentic
8.5% max. (K.F.)
215.0°C to 218.0°C
95%
94% min. (HPLC)
MFCD00150807
Solubility in water: 200g/L (20°C)
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
527.12
527.12
95%
FDS