Learn More
BioTek™ Cytation™ 5 Cell Imaging Multi-Mode Reader PROMO
Used in Automated digital microscopy and imaging. Cytation™ Cell Imaging Multi-Mode Reader provides live cell assay support with environmental controls.
Brand:  BioTek™ CYT5MFAVPROMOQ22016
Additional Details : Weight : 36.30000kg
 View more versions of this product
Product Code. 15638645
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Features:
  • Automated digital microscopy
  • Brightfield, colour brightfield, phase contrast, and fluorescence imaging
  • Live cell assay support with environmental controls
  • Dual reagent injector compatible
  • Modular and upgradable to add multimode detection
  • BioSpa 8 automated incubator compatible to automate live cell workflows
Laser autofocus offers:
  • Improved speed with dim fluorophores and transmitted light
  • Scan 96 wells at 4x with DAPI in less than 3 minutes
  • Greater reproducibility with no effects from well debris
  • Accurate in long term time-lapseworkflows
  • Minimised photobleaching with lower phototoxicity for live cell assays
Gen5 Image+ software provides:
  • Quick and easy cell counting
  • Subpopulation analysis
  • Automated movie maker
  • Image and graph annotations
Specifications
Specifications
Cell Imaging Multi-Mode Reader
Monochromator: 6-well to 384-well plates, Filters: 6-well to 1536-well plates, Imaging: 6-well to 1536-well plates
Linear, orbital, double orbital
Gen5™ Microplate Reader and Imager Software
CYT5MFAV
0 to 20%
Hybrid technology
4-Zone incubation to 65°C
Laser Autofocus and Gen5 Image+ Software
Fluorescence, brightfield, color brightfield, monochromator and filter based fluorescence, luminiscence and alpha detection
Endpoint, kinetic, spectral scanning, well area scanning
1 to 19%
Videos