missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin/Stefin Homolog 2 Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Cystatin/Stefin Homolog 2 |
---|---|
Clone | 237305 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Cystatin/Stefin Homolog 2 Monoclonal specifically detects Cystatin/Stefin Homolog 2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin/Stefin Homolog 2 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
NP_084009 | |
76770 | |
E. coli-derived recombinant mouse Cystatin/Stefin Homolog 2 Met2-Phe97 Accession # NP_084009 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cystatin/Stefin Homolog 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, S, SA, SN, recombinant mouse Cystain B, C, E/M, rhFetuin A, rhFetuin B, rhHPRG, or rhKininostatin is observed. |
237305 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
010005H15Rik, Csta2, Cystatin H2 | |
2010005H15Rik | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only