missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin S Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Cystatin S |
---|---|
Clone | 201805 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Cystatin S Monoclonal specifically detects Cystatin S in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin S | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P01036 | |
1472 | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin S Ser21-Ala141 Accession # P01036 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Cystatin S in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin SA is observed and no cross-reactivity with rhCystatin D, E, SN, rhFetuin A, rhFetuin B, or rhHPRG (Histidine-Proline-Rich Glycoprotein) is observed. In Western blots, approximately 15% cross-reactivity with rhCystatins SA and SN (under reducing conditions only) is observed and no cross-reactivity with rhCystatin A, B, C, D, E/M, rhFetuin A, rhFetuin B, or rhHPRG is observed. |
201805 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CST4, cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1 | |
CST4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only