missing translation for 'onlineSavingsMsg'
Learn More

Cystamine dihydrochloride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7026711
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
56-17-7
C4H14Cl2N2S2
225.19
MFCD00012905
YUFRRMZSSPQMOS-UHFFFAOYSA-N
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
5941
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
C(CSSCCN)N.Cl.Cl
This item is not returnable. View return policy
56-17-7
C4H14Cl2N2S2
225.19
MFCD00012905
YUFRRMZSSPQMOS-UHFFFAOYSA-N
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
5941
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
C(CSSCCN)N.Cl.Cl

Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 217°C to 220°C
Assay Percent Range 98+%
Linear Formula (H2NCH2CH2S)2·2HCl
Quantity 25 g
Beilstein 3616850
Sensitivity Hygroscopic
Merck Index 14,2776
Solubility Information Soluble in water.
Formula Weight 225.2
Percent Purity ≥97%
Chemical Name or Material Cystamine dihydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'