missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549351
Additional Details : Weight : 0.01000kg
Description
CYS1 Polyclonal antibody specifically detects CYS1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CYS1 | |
Polyclonal | |
Unconjugated | |
CYS1 | |
2900006B19Rik; AV218859; Ccap; cilia-associated protein; ck; congenital polycystic kidneys; cpk; Cys1; cystin 1; cystin cilia associated protein; cystin-1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
12879, 192668 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q717R9, Q8R4T1 | |
CYS1 | |
This CYS1 antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide with amino acid positions 128-158 of human CYS1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |