missing translation for 'onlineSavingsMsg'
Learn More

CyPPA, Tocris Bioscience™

Activator of KCa2.2 and KCa2.3 channels

Brand:  Tocris (Bio-Techne) 295350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442124

  • £570.72 / 50mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Activator of small conductance Ca2+-activated K+ channels that displays selectivity for KCa2.2 (SK2) and KCa2.3 (SK3) channels (EC50 values are 5.6 and 14 μM for KCa2.3 and KCa2.2 respectively). Displays no activity at KCa2.1 (SK1) and KCa3.1 (IK) channels.
Specifications

Chemical Identifiers

73029-73-9
285.395
cyppa, n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methylpyrimidin-4-amine, cyclohexyl-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-pyrimidin-4-yl-amine, n-cyclohexyl-2-3,5-dimethylpyrazol-1-yl-6-methylpyrimidin-4-amine, n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine, d00dif, cyppa hplc , solid, 2-3,5-dimethyl-1h-pyrazole-1-yl-4-methyl-6-cyclohexylamino pyrimidine, n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methyl-4-pyrimidinamine, n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine
N-cyclohexyl-2-(3,5-dimethylpyrazol-1-yl)-6-methylpyrimidin-4-amine
C16H23N5
USEMRPYUFJNFQN-UHFFFAOYSA-N
909822
CC1=CC(=NC(=N1)N2C(=CC(=N2)C)C)NC3CCCCC3

Specifications

CyPPA
73029-73-9
cyppa, n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methylpyrimidin-4-amine, cyclohexyl-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-pyrimidin-4-yl-amine, n-cyclohexyl-2-3,5-dimethylpyrazol-1-yl-6-methylpyrimidin-4-amine, n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine, d00dif, cyppa hplc , solid, 2-3,5-dimethyl-1h-pyrazole-1-yl-4-methyl-6-cyclohexylamino pyrimidine, n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methyl-4-pyrimidinamine, n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine
CC1=CC(=NC(=N1)N2C(=CC(=N2)C)C)NC3CCCCC3
285.395
285.39
50mg
C16H23N5
USEMRPYUFJNFQN-UHFFFAOYSA-N
N-cyclohexyl-2-(3,5-dimethylpyrazol-1-yl)-6-methylpyrimidin-4-amine
909822
>99%
Videos