Learn More
Tocris Bioscience™ CYM 5442 hydrochloride
Selective S1P1 receptor agonist
Brand:  Tocris Bioscience™ 360110
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443274
  • £219.03 / 10mg
Estimated Shipment date
from Supplier 20-12-2019
Description
Description
Potent and selective S1P1 agonist in vitro (EC50 = 1.35 nM). Activates S1P1-mediated p42/p44 MAPK phosphorylation in CHO-K1 cells transfected with S1P1. Induces acute lymphopenia in mice. Brain penetrant.
Specifications
Specifications
1783987-80-3
CYM 5442 hydrochloride
445.94
CYM 5442 hydrochloride
>98