missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Cylindrical PTFE Stir Bars

Provides stable stirring action. Cowie Technology™ Cylindrical PTFE Stir Bars are a popular general-purpose stirrer for a wide variety of applications.

Brand:  Cowie Technology™ 001.125

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 10289701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications

Specifications

6 mm
White
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Cylinder
PTFE encapsulated Alnico V magnet core
5/Pk.
Yes
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers