missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Cylindrical PTFE Stir Bars

Provides stable stirring action. Cowie Technology™ Cylindrical PTFE Stir Bars are a popular general-purpose stirrer for a wide variety of applications.

Brand:  Cowie Technology™ 001.130.8

Code : NEW

Additional Details : Weight : 0.00520kg

 View more versions of this product

Product Code. 10425832

  • £3.50 / Each


In Stock 6
Add to basket

Description

Description

  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
5/Pk.
8mm
Cylindrical
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
Containers & Vessels / Stirrer Hot Plates
PTFE Stir Bars Cylindrical
30mm
Videos