Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04375814
Code : 01
Additional Details : Weight : 1.00000kg
Description
- With scale (approx. volume/mL)
- With pressure equalizing tube
- With screwthread retaining device
- Cylindrical shape with NS socket and cone
- Corrosion-free and resistant to thermal shock
Specifications
Pressure Equalizing Tube | |
10mL | |
4.0mm bore | |
2 | |
500 mL |
Yes | |
29/32 NS | |
Dropping funnel | |
PTFE | |
Borosilicate Glass |