Learn More
Kartell™ Polypropylene Centrifuge Tubes
Manufactured from highly translucent polypropylene for  enhanced visibility and good chemical resistance. Autoclavable, rimmed Kartell™ Cylindrical Centrifuge Tubes deliver stabilitiy in a properly balanced rotor and are suitable for use as a conventional test tube.
Brand:  Kartell™ 305
Code : 02
Additional Details : Weight : 0.00400kg
Product Code. 11371235
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Solid polypropylene construction translates to safety and economy with reduced breakage in the lab
  • Withstands temperatures up to 120°C
  • Rimmed
  • Autoclavable
  • Doubles as a laboratory test tube*
*Cannot be used with naked flame
Videos
Documentation