missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
124.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Flash Point | −11°C |
Packaging | AcroSeal™ Glass bottle |
Linear Formula | C3H5MgBr |
Quantity | 100 mL |
Solubility Information | Solubility in water: reacts |
Formula Weight | 145.28 |
Chemical Name or Material | Cyclopropylmagnesium bromide |
Safety and Handling
Product Identifier
- Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'