missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
41.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.1400g/mL |
Boiling Point | 60.0°C to 61.0°C (29.0 mmHg) |
Flash Point | 93°C |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 1 g |
Specific Gravity | 1.14 |
Formula Weight | 100.12 |
Percent Purity | 97% |
Chemical Name or Material | Cyclopropylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'