missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 97.0 |
Density | 1.07g/mL |
Assay Percent Range | 97% |
Linear Formula | C3H5NCS |
Quantity | 5 g |
Formula Weight | 99.16 |
Percent Purity | 97% |
Chemical Name or Material | Cyclopropyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'