missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Isothiocyanate, 97%, ACROS Organics™

Brand:  Acros Organics 375490050

Code : 29

Additional Details : CAS Number : 56601-42-4 Weight : 0.05500kg Transport : UN number : 3286 Chem class : 3 Pack group : II


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10270393

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Cyclopropyl isothiocyanate
97%
C3H5NCS
MFCD00040874
≥96.0%
C4H5NS
99.16

Specifications

Cyclopropyl isothiocyanate
Clear colorless to slightly yellow
56601-42-4
Glass bottle
≥96.0%
C4H5NS
99.16
1.0700g/mL
5g
Authentic
Liquid
97%
C3H5NCS
MFCD00040874
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclopropyl isothiocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS