missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Carbinol, 99%, ACROS Organics™

Brand:  Acros Organics 296822500

Code : 29

Additional Details : CAS Number : 2516-33-8 Weight : 0.27250kg Transport : UN number : 2920 Chem class : 1b Pack group : II

Quantity 250mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10531794

  • POA / 250mL

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

Cyclopropyl carbinol
99%
C3H5CH2OH
MFCD00001309
≥98.5%
C4H8O
72.11
Cyclopropanemethanol

Specifications

Cyclopropyl carbinol
Clear colorless
Clear colorless liquid
Authentic
Liquid
99%
C4H8O
72.11
Cyclopropanemethanol
0.8900g/mL
250mL
2516-33-8
Glass bottle
≥98.5%
1.4310 to 1.4330 (20°C, 589nm)
C3H5CH2OH
MFCD00001309
Videos
SDS