missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7081222
17.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5500-21-0
C4H5N
67.091
MFCD00001269
AUQDITHEDVOTCU-UHFFFAOYSA-N
cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
79637
cyclopropanecarbonitrile
C1CC1C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5500-21-0
C4H5N
67.091
MFCD00001269
AUQDITHEDVOTCU-UHFFFAOYSA-N
cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
79637
cyclopropanecarbonitrile
C1CC1C#N

Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Density 0.905
Boiling Point 134°C to 135°C
Flash Point 32°C (89°F)
Odor Recognizable
Refractive Index 1.421
Quantity 10 g
UN Number UN3275
Beilstein 635790
Solubility Information Miscible with water.
Formula Weight 67.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarbonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'