missing translation for 'onlineSavingsMsg'
Learn More

Cyclopiazonic acid, Tocris Bioscience™

Inhibitor of SERCA ATPase

Brand:  Tocris (Bio-Techne) 123510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13428933

  • £152.88 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
Specifications

Chemical Identifiers

18172-33-3
336.391
cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
C20H20N2O3
SZINUGQCTHLQAZ-AFAVFJNCSA-N
54711281

Specifications

10mg
18172-33-3
cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
54711281
>98%
Cyclopiazonic acid
C20H20N2O3
SZINUGQCTHLQAZ-AFAVFJNCSA-N
336.391
336.39
Videos
Documents