Learn More
Cyclophilin C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558679
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VFGKVIDGMT VVHSIELQAT DGHDRPLTNC SIINSGKIDV KTPFVVEIAD W Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
Cyclophilin C Polyclonal antibody specifically detects Cyclophilin C in Human samples. It is validated for Immunohistochemistry.Specifications
Cyclophilin C | |
Polyclonal | |
Unconjugated | |
Ppic | |
Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P45877 | |
Ppic | |
Recombinant protein corresponding to Human Cyclophilin C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only