missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene, 98+%, Thermo Scientific Chemicals

Catalog Number 11324198
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
142-29-0
C5H8
68.119
MFCD00001394
LPIQUOYDBNQMRZ-UHFFFAOYSA-N
1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
8882
CHEBI:49155
cyclopentene
C1CC=CC1
This item is not returnable. View return policy
142-29-0
C5H8
68.119
MFCD00001394
LPIQUOYDBNQMRZ-UHFFFAOYSA-N
1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
8882
CHEBI:49155
cyclopentene
C1CC=CC1

Cyclopentene is used in organic synthesis for cross-linking resin. It is used as an intermediate in Industries such as Agrochemical, Dyestuff, Pharmaceutical, syntheses material, Chemical. It is used as a monomer for synthesis of plastics and rubber. It is also used for the synthesis of various chemicals, such as 1,1,2-Trimethylcyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -135°C
Color Colorless
Density 0.773 g/mL
Boiling Point 44°C to 45°C
Flash Point -29°C (-20°F)
Refractive Index 1.4225
Quantity 100 mL
UN Number UN2246
Beilstein 635707
Sensitivity Air Sensitive
Solubility Information It is soluble in most organic solvents like ethanol and ether. It is immiscible with water.
Formula Weight 68.12
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Cyclopentene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'