missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, 98%, Pure, ACROS Organics™

Brand:  Acros Organics 111481000

Code : 29

Additional Details : CAS Number : 287-92-3 Weight : 0.12500kg Transport : UN number : 1146 Chem class : 3 Pack group : II

Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10638143

  • £22.20 / 100mL


EU Stock 10
Add to basket

Specifications

Chemical Identifiers

Cyclopentane
Extra Pure
98%
C5H10
MFCD00001356
287-92-3
98%
C5H10
70.15

Specifications

Cyclopentane
−7°C
White
100mL
Authentic
1.4050 to 1.4070
0.01 % max.
98%
C5H10
70.15
05, 19
0.7500g/mL
50.0°C
− 94.0°C
287-92-3
Glass bottle
Extra Pure
Powder
98%
C5H10
MFCD00001356
15, 2736
Videos
SDS