missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclopentadienylvanadium tetracarbonyl, 97+%

Brand:  Alfa Aesar™ 081123.04

Code : NEW

Additional Details : CAS Number : 12108-04-2 Weight : 0.00200kg Transport : UN number : 3466 Chem class : 6.1 Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11304929

  • £280.30 / 2g


Estimated Shipment date
from Supplier 19-07-2018
Add to basket

Specifications

Chemical Identifiers

Cyclopentadienylvanadium tetracarbonyl
C9H5O4V
MFCD00014259
Tetracarbonylcyclopentadienylvanadium
12108-04-2
228.08
UN3466

Specifications

Cyclopentadienylvanadium tetracarbonyl
80° to 100°C (0.1mmHg) (subl.)
2g
Crystalline
C9H5O4V
MFCD00014259
Tetracarbonylcyclopentadienylvanadium
Air Sensitive
138°C
12108-04-2
≥97%
228.08
UN3466
Videos
SDS