missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals

Catalog Number p-97290
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1270-98-0
C5H5Cl3Ti
219.36
MFCD00003724
QOXHZZQZTIGPEV-UHFFFAOYSA-K
This item is not returnable. View return policy
1270-98-0
C5H5Cl3Ti
219.36
MFCD00003724
QOXHZZQZTIGPEV-UHFFFAOYSA-K

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 170°C
Color Yellow to Orange
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 g
Assay Percent Range 99%
Solubility Information Solubility in water: reacts
Formula Weight 219.36
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cyclopentadienyltitanium trichloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentadienyltitanium trichloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'