missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylrhenium tricarbonyl, 99%, Thermo Scientific™

Catalog Number p-7050050
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
12079-73-1
C8H8O3Re
338.36
MFCD00058885
NEXLWSRPYZNSNH-UHFFFAOYSA-N
cyclopentadienylrhenium tricarbonyl, rhenium, tricarbonyl eta5-2,4-cyclopentadien-1-yl
This item is not returnable. View return policy
12079-73-1
C8H8O3Re
338.36
MFCD00058885
NEXLWSRPYZNSNH-UHFFFAOYSA-N
cyclopentadienylrhenium tricarbonyl, rhenium, tricarbonyl eta5-2,4-cyclopentadien-1-yl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.5 g
UN Number UN3466
Solubility Information Insoluble in water.
Formula Weight 335.33
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Cyclopentadienylrhenium tricarbonyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'