missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7051131
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
62927-99-5
MFCD00269826
Tetrachlorocyclopentadienylmolybdenum(V)
This item is not returnable. View return policy
62927-99-5
MFCD00269826
Tetrachlorocyclopentadienylmolybdenum(V)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Purple-Red
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 302.85
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Cyclopentadienylmolybdenum(V) tetrachloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'