missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienyliron dicarbonyl dimer, 99%, ACROS Organics™

Brand:  Acros Organics 209301000

Code : 29

Additional Details : CAS Number : 12154-95-9 Weight : 0.15000kg Transport : UN number : 3466 Chem class : 1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10677094

  • £212.60 / 100g


Estimated Shipment
05-03-2018
Add to basket


Specifications

Chemical Identifiers

Cyclopentadienyliron dicarbonyl dimer
C14H10Fe2O4
MFCD00001436
99%
353.92

Specifications

Cyclopentadienyliron dicarbonyl dimer
194.0°C
12154-95-9
Authentic
Crystalline Powder
99%
353.92
Red-purple to very dark brown
100g
98.5% min.
Glass bottle
99%
C14H10Fe2O4
MFCD00001436
Videos
SDS