missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (Cyclooctatetraene)iron tricarbonyl, 98%

Brand:  Alfa Aesar™ 039402.04

Code : NEW

Additional Details : CAS Number : 12093-05-9 Weight : 0.00200kg Transport : UN number : 3466 Chem class : 1 Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11308188

  • £265.90 / 2g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

(Cyclooctatetraene)iron tricarbonyl
244.03
UN3466
12093-05-9
MFCD00001755
Tricarbonylcyclooctatetraene iron

Specifications

(Cyclooctatetraene)iron tricarbonyl
Crystalline
244.03
UN3466
Red-brown
2g
12093-05-9
98%
MFCD00001755
Tricarbonylcyclooctatetraene iron
93° to 95°C
Air Sensitive
Videos
SDS