missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylacetylene, 98%, Thermo Scientific Chemicals

Catalog Number p-3523409
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
931-48-6
C8H12
108.18
MFCD00001513
SSDZYLQUYMOSAK-UHFFFAOYSA-N
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
70263
ethynylcyclohexane
C#CC1CCCCC1
This item is not returnable. View return policy
931-48-6
C8H12
108.18
MFCD00001513
SSDZYLQUYMOSAK-UHFFFAOYSA-N
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
70263
ethynylcyclohexane
C#CC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 0.8280g/mL
Boiling Point 130°C to 132°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11C∽CH
Refractive Index 1.4530 to 1.455
Quantity 1 g
Specific Gravity 0.828
Formula Weight 108.18
Percent Purity 98%
Physical Form Solution
Chemical Name or Material Cyclohexylacetylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclohexylacetylene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'