missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclohexyl methyl methylphosphonate, 99%

Brand:  Alfa Aesar™ 044426.04

Code : NEW

Additional Details : CAS Number : 7040-52-0 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11410720

  • £133.80 / 2g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Cyclohexyl methyl methylphosphonate
CH3P(O)(OCH3)(OC6H11)
MFCD08064211
7040-52-0
192.09

Specifications

Cyclohexyl methyl methylphosphonate
7040-52-0
CH3P(O)(OCH3)(OC6H11)
MFCD08064211
2g
99%
192.09
Videos
SDS