missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl ketone, 95%, Thermo Scientific Chemicals

Catalog Number p-7025502
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
823-76-7
C8H14O
126.199
MFCD00040418
RIFKADJTWUGDOV-UHFFFAOYSA-N
1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
13207
1-cyclohexylethanone
CC(=O)C1CCCCC1
This item is not returnable. View return policy
823-76-7
C8H14O
126.199
MFCD00040418
RIFKADJTWUGDOV-UHFFFAOYSA-N
1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
13207
1-cyclohexylethanone
CC(=O)C1CCCCC1

It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point -34°C
Density 0.917
Boiling Point 181°C to 183°C
Flash Point 66°C (151°F)
Assay Percent Range 0.95
Refractive Index 1.452
Quantity 5 g
Beilstein 507229
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 126.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclohexyl methyl ketone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'