missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number 11324539
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1122-82-3
C7H11NS
141.232
MFCD00003841
MZSJGCPBOVTKHR-UHFFFAOYSA-N
cyclohexyl isothiocyanate, cyclohexane, isothiocyanato, isothiocyanocyclohexane, cyclohexyl-isothiocyanat, isothiocyanic acid, cyclohexyl ester, cyclohexylisothiocyanate, cyclohexyl isothiocyanate, isothiocyanato, cyclohexyl-isothiocyanat german, isothiocyanato-cyclohexane, isothiocyanic acid cyclohexyl ester
14289
isothiocyanatocyclohexane
C1CCC(CC1)N=C=S
This item is not returnable. View return policy
1122-82-3
C7H11NS
141.232
MFCD00003841
MZSJGCPBOVTKHR-UHFFFAOYSA-N
cyclohexyl isothiocyanate, cyclohexane, isothiocyanato, isothiocyanocyclohexane, cyclohexyl-isothiocyanat, isothiocyanic acid, cyclohexyl ester, cyclohexylisothiocyanate, cyclohexyl isothiocyanate, isothiocyanato, cyclohexyl-isothiocyanat german, isothiocyanato-cyclohexane, isothiocyanic acid cyclohexyl ester
14289
isothiocyanatocyclohexane
C1CCC(CC1)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.033g/mL
Boiling Point 219°C
Odor Pungent
Refractive Index 1.5380
Quantity 5 g
UN Number UN2922
Beilstein 774525
Sensitivity Moisture Sensitive
Formula Weight 141.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'