missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Isocyanate, 98+%, ACROS Organics™

Brand:  Acros Organics 111310025

Code : 29

Additional Details : CAS Number : 3173-53-3 Weight : 4.00000kg Transport : UN number : 2488 Chem class : 1 Pack group : I


Quantity 2.5kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10452835

  • £198.00 / 2.50kg


Estimated Shipment
30-05-2018
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Cyclohexyl isocyanate
95-50-1
98+%
C6H11NCO
MFCD00003840
98+%
≥98.0%
C7H11NO
125.17

Specifications

Cyclohexyl isocyanate
Clear colorless to light amber
total other isocyanates (GC): < 2.0%
95-50-1
Authentic
1.4540 to 1.4560 (20°C, 589nm)
Product may develop some turbidity or precipitate
98+%
C6H11NCO
MFCD00003840
0.9800g/mL
2.5kg
98+%
hydrolyzable chloride (titr.): < 0.3%
Glass bottle
Liquid
≥98.0%
C7H11NO
125.17
12, II, 12
Videos
SDS