missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Isocyanate, 98+%, Thermo Scientific Chemicals

Catalog Number 10648162
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3173-53-3, 95-50-1
C7H11NO
125.17
MFCD00003840
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
18502
CHEBI:136604
isocyanatocyclohexane
O=C=NC1CCCCC1
This item is not returnable. View return policy
3173-53-3, 95-50-1
C7H11NO
125.17
MFCD00003840
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
18502
CHEBI:136604
isocyanatocyclohexane
O=C=NC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Amber to Colorless
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H11NCO
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Quantity 5 g
Beilstein 12, II, 12
Formula Weight 125.17
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material Cyclohexyl isocyanate, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'