missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene Sulfide (Tech.), ca 85%, ACROS Organics™

Brand:  Acros Organics 111220100

Code : 29

Additional Details : CAS Number : 286-28-2 Weight : 0.06000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10224330

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Cyclohexene sulfide
286-28-2
≥80%
C6H10S
MFCD00005163
tech., ca. 85%
Technical Grade
ca. 85%
114.2
7-Thiabicyclo[4.1.0]heptane

Specifications

Cyclohexene sulfide
286-28-2
Technical Grade
≥80%
Glass bottle
114.2
7-Thiabicyclo[4.1.0]heptane
10g
tech., ca. 85%
Authentic
Opalescent Liquid
ca. 85%
C6H10S
MFCD00005163
Yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclohexene sulfide

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS