missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, 99+%, Thermo Scientific Chemicals

Catalog Number p-3345827
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
108-94-1
C6H10O
98.145
MFCD00001625
JHIVVAPYMSGYDF-UHFFFAOYSA-N
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
7967
CHEBI:17854
cyclohexanone
C1CCC(=O)CC1
This item is not returnable. View return policy
108-94-1
C6H10O
98.145
MFCD00001625
JHIVVAPYMSGYDF-UHFFFAOYSA-N
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
7967
CHEBI:17854
cyclohexanone
C1CCC(=O)CC1

Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

Solubility
Miscible with diethyl ether, acetone and alcohol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, red metals, lead, amines, sulfuric acid, nitric acid, strong acids, aliphatic amines, rubber, resins and alkalies.
TRUSTED_SUSTAINABILITY
Melting Point °C to 26°C
Density 0.9478
Boiling Point 154°C to 156°C
Quantity 500 mL
Flash Point 47°C (116°F)
Physical Form Liquid
Refractive Index 1.4507
Assay Percent Range ≥99%
UN Number UN1915
Beilstein 385735
Merck Index 14,2726
Solubility Information Miscible with diethyl ether,acetone and alcohol. Slightly miscible with water.
Formula Weight 98.15
Percent Purity ≥99%
Chemical Name or Material Cyclohexanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'