missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclohexano-15-crown-5, cis + trans, 97%

Brand:  Alfa Aesar™ L16964.03

Code : NEW

Additional Details : CAS Number : 17454-48-7 Weight : 0.00100kg

Quantity 1g

Product Code. 15479049

  • £46.30 / 1g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Specifications

Cyclohexano-15-crown-5
180°C (0.4mmHg)
Cis + Trans
97%
274.36
1682984
>100°C (212°F)
1g
17454-48-7
C14H26O5
MFCD00054545
2,3-Cyclohexano-1,4,7,10,13-pentaoxacyclopentadecane
Videos
SDS